Ammonium iron(II) sulphate 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®

Leverandør: Merck
Danger

Synonymer: di-Ammonium iron (II) sulphate, Ammonium ferrous sulphate

M-Clarity™ quality level = MQ100

1.09864.0001
1.09864.0001EA 1070 NOK
1.09864.0001
Ammonium iron(II) sulphate 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Ammonium iron(II) sulphate
Formel: H₈FeN₂O₈S₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00051039
CAS-nummer: 10045-89-3
EINECS: 233-151-8
FN: 2796
ADR: 8,

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Density 1.25 g/cm3 (20 °C)
pH 0.4 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR