Natriumhydroksid 5 mol/l (5 N) in aqueous solution, Titrisol®, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ100

1.09913.0001
1.09913.0001EA 602 NOK
1.09913.0001
Natriumhydroksid 5 mol/l (5 N) in aqueous solution, Titrisol®, Supelco®
Natriumhydroksid
Formel: NaOH
Kokepunkt: 109 °C (1013 hPa)
Tetthet: ≤1,11 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
FN: 1824
ADR: 8,

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Density 1.185 g/cm3 (20 °C)
pH 14 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR