Kaliumdikromat 1/60 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Leverandør: Merck
Danger
M-Clarity™ quality level = MQ100
1.09928.0001
1.09928.0001EA 354 NOK
1.09928.0001
Kaliumdikromat 1/60 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Kaliumdikromat
Formel: K₂Cr₂O₇
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011367
CAS-nummer: 7778-50-9
EINECS: 231-906-6
FN: 3287
ADR: 6.1,
Bestill nå
Spesifikasjon Testresultater
Solubility (20 °C) soluble
Density 1.06 g/cm3 (20 °C)
pH 4.1 (H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic materials