Natriumtiosulfat 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ100

1.09950.0001 1.09950.0050
1.09950.0001EA 350 NOK
1.09950.0001 1.09950.0050
Natriumtiosulfat 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Natriumtiosulfat
Formel: Na₂S₂O₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003499
CAS-nummer: 7772-98-7
EINECS: 231-867-5

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Density 1.22 g/cm3 (20 °C)
pH 9 - 10 (H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR