Natriumhydroksid 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ100

1.09959.0001 1.09959.0050
1.09959.0001EA 385 NOK
1.09959.0001 1.09959.0050
Natriumhydroksid 0.1 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Natriumhydroksid
Formel: NaOH
Kokepunkt: ∼105 °C (1013 hPa)
Tetthet: 1,327 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
FN: 1824
ADR: 8,

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Density 1.09 g/cm3 (20 °C)
pH 13.8 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR