Natriumhydroksid 0.01 mol/l aqueous solution_concentrated, Supelco®

Leverandør: Merck
Warning

Synonymer: Caustic soda

MQ100

1.09961.0001
1.09961.0001EA 375 NOK
1.09961.0001
Natriumhydroksid 0.01 mol/l aqueous solution_concentrated, Supelco®
Natriumhydroksid
Formel: NaOH
MW: 40 g/mol
Kokepunkt: ∼105 °C (1013 hPa)
Smeltepunkt: 323
Tetthet: 1.327 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
FN: 1824
ADR: 8
Merck Indeks: 14,08627

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Density 1.01 g/cm3 (20 °C)
pH 13 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR