Saltsyre 0.5 mol/l concentrated aqueous solution, Titrisol®, Supelco®

Leverandør: Merck
Warning

Synonymer: Hydrochloric acid solution, Muriatic acid

M-Clarity™ quality level = MQ100

1.09971.0001
1.09971.0001EA 211.33 NOK
1.09971.0001
Saltsyre 0.5 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Saltsyre
Formel: HCl
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
FN: 1789
ADR: 8,

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Density 1.1 g/cm3 (20 °C)
pH  - 1 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR