Svovelsyre 0.5 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Leverandør: Merck
Danger
M-Clarity™ quality level = MQ100
1.09981.0001
1.09981.0001EA 307.5 NOK
1.09981.0001
Svovelsyre 0.5 mol/l concentrated aqueous solution, Titrisol®, Supelco®
Svovelsyre
Formel: H₂SO₄
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064589
CAS-nummer: 7664-93-9
EINECS: 231-639-5
FN: 2796
ADR: 8,
Limited Time Sale
Select items below are currently on sale
Bestill nå
Spesifikasjon Testresultater
Solubility (20 °C) soluble,(caution ! development of heat)
Boiling Point 135 °C
Density 1.29 g/cm3 (20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material