Svovelsyre 0.005 mol/l concentrated aqueous solution, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ100

1.09982.0001
1.09982.0001EA 326 NOK
1.09982.0001
Svovelsyre 0.005 mol/l concentrated aqueous solution, Supelco®
Svovelsyre
Formel: H₂SO₄
Tetthet: 1,01 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064589
CAS-nummer: 7664-93-9
EINECS: 231-639-5
FN: 3264
ADR: 8,

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Density 1.01 g/cm3 (20 °C)
pH 1.1 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR