Titriplex® III concentrate 0.1 M, Titrisol®, Supelco®

Leverandør: Merck
Warning

Synonymer: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt

M-Clarity™ quality level = MQ100

1.09992.0001
1.09992.0001EA 257.73 NOK
1.09992.0001
Titriplex® III concentrate 0.1 M, Titrisol®, Supelco®
EDTA dinatrium salt

Concentration after dilution to 1 liter: c(C₁₀H₁₄N₂Na₂O₈ ∙ 2 H₂O) = 0.1 mol/l

Formel: C₁₀H₁₄N₂Na₂O₈
Tetthet: 1,14 g/cm3 (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00070672
CAS-nummer: 139-33-3
EINECS: 205-358-3

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Density 1.14 g/cm3 (20 °C)
pH 7.4 - 7.6 (H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR