Vanadium concentrate, 1000 mg V in 8,6% sulphuric acid (fra VO(SO₄)) Titrisol® standard til AAS, Supelco®
Leverandør: Merck
1.09994.0001
1.09994.0001EA 665 NOK
1.09994.0001
Vanadium concentrate, 1000 mg V in 8,6% sulphuric acid (fra VO(SO₄)) Titrisol® standard til AAS, Supelco®
Standarder Kationstandarder Vanadium Standards
Sertifiseringer: Traceable to SRM from NIST