Dichloroisocyanuric acid sodium salt dihydrate, Supelco®

Leverandør: Merck
Warning

Synonymer: NaDCC

M-Clarity™ quality level = MQ100

1.10888.0100
1.10888.0100EA 3260 NOK
1.10888.0100
Dichloroisocyanuric acid sodium salt dihydrate, Supelco®
Dichloroisocyanuric acid sodium salt dihydrate
Formel: C₃Cl₂N₃NaO₃·2H₂O
MW: 255.98 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149404
CAS-nummer: 51580-86-0
EINECS: 220-767-7
FN: 3077
ADR: 9

Bestill nå

Spesifikasjon Testresultater

Solubility 250 g/l (25 °C)
Melting Point 240 - 250 °C (decomposition)
Molar Mass 255.98 g/mol
Bulk Density 980 kg/m3
pH 6.7 (10 g/l, H2O, 20 °C)
Hazard Class 9
Hazard Class Description Miscellaneous hazardous material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR