Natriumhydroksid 4 mol/l (4 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Leverandør: Merck
Danger
Synonymer: Caustic soda
M-Clarity™ quality level = MQ100
1.11584.5000
1.11584.5000EA 941 NOK
1.11584.5000
Natriumhydroksid 4 mol/l (4 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Natriumhydroksid
Formel: NaOH
MW: 40 g/mol
Kokepunkt: 1390
Smeltepunkt: 323
Tetthet: ≤1.11 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
FN: 1824
ADR: 8
Merck Indeks: 14,08627
Bestill nå
Spesifikasjon Testresultater
Form liquid
Amount-of-substance concentration 3,980-4,020
Measurement uncertainty + / - 0,012
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C.