Hemacolor® Rapid staining of blood smear kit stain for hematology, for histologi, Sigma-Aldrich®
Leverandør: Merck
Danger
1.11661.0001 1.11674.0001
1.11661.0001EA 2225 NOK
1.11661.0001 1.11674.0001
Hemacolor® Rapid staining of blood smear kit stain for hematology, for histologi, Sigma-Aldrich®
Hemacolor® Rapid staining of blood smear
For use in hematology and clinical cytology
The Hemacolor® staining kit combines the brilliance and reproducibility of the Pappenheim staining with the time-advantage of a rapid stain. The Hemacolor® staining kit contains a fixing solution, one red (eosin) and one blue (azur) staining solution and phosphate buffer tablets pH 7.2 acc. to Weise. The staining solutions are separately applied, which results in a high degree of stability. The conventionally appearing precipitates in the hematological staining solutions, could be avoided in this kit. In addition to blood and bone marrow smears, Hemacolor® can be used in clinical cytology, e.g. for staining urine, sputum, FNAB, sperm, effusions and lavages. A kit contains safe 100-ml or 500-ml PE-bottles and six buffer tablets; sufficient for staining 250 and up to 1000 slides. The Hemacolor® staining kit is certified and registered as CE / IVD product for applications in diagnostics.
This product is a kit which contains the following:
Hemacolor® Rapid staining of blood smear solution 1, fixing solution (Cat no. 111955)
Hemacolor® Rapid staining of blood smear solution 2, colour reagent red (Cat no. 111956)
Hemacolor® Rapid staining of blood smear solution 3, colour reagent blue (Cat no. 111957)
Lagringstemperatur: Romtemperatur
FN: 1230
ADR: 3 (6.1),
IVDLogo
Bestill nå
Spesifikasjon Testresultater
Suitability for microscopy (Blood smear) passes test
Erythrocytes pink to brownish
Nuclei red-violet
Eosinophilic granules red to red-brown
Neutrophilic granules light violet
Lymphocyte cytoplasm Blue