Phenol red sodium salt, Supelco®

Leverandør: Merck

Synonymer: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt, Phenol red water soluble, Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bisphenolate S,S-dioxide

MQ200

1.11748.0005
1.11748.0005EA 692 NOK
1.11748.0005
Phenol red sodium salt, Supelco®
Phenol red sodium salt
Formel: C₁₉H₁₃NaO₅S
MW: 376.35 g/mol
Smeltepunkt: 285
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00066901
CAS-nummer: 34487-61-1
EINECS: 252-057-8
Merck Indeks: 12,07397

Bestill nå

Spesifikasjon Testresultater

Solubility 100 g/l (20 °C)
Melting Point 285 °C
Molar Mass 376.35 g/mol

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR