Sodium octyl sulphate, Sigma-Aldrich®

Leverandør: Merck


M-Clarity™ quality level = MQ200

1.12268.0005 1.12268.0500
1.12268.0005EA 2000 NOK
1.12268.0005 1.12268.0500
Sodium octyl sulphate, Sigma-Aldrich®
Sodium octyl sulphate
Formel: C₈H₁₇NaO₄S
Smeltepunkt: 181…183 °C
Tetthet: 1,266 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007470
CAS-nummer: 142-31-4
EINECS: 205-535-5

Bestill nå

Spesifikasjon Testresultater

Hill Formula C₈H₁₇NaO₄S
Chemical formula C₈H₁₇NaSO₄
Molar Mass 232.26 g/mol
Melting Point 195 °C (decomposition)
pH value 6 - 8 (10 g/l, H₂O, 20 °C)
Bulk density 470 kg/m³
Solubility (20 °C) soluble
LD 50 oral LD50 Rat 3200 mg/kg
Storage class 10 - 13 Other liquids and solids
WGK WGK 2 water endangering


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR