Nikkel, pulver, Sigma-Aldrich®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ200

1.12277.0250 1.12277.1000
1.12277.0250EA 1540 NOK
1.12277.0250 1.12277.1000
Nikkel, pulver, Sigma-Aldrich®
Nikkel
Formel: Ni
Kokepunkt: 2371 °C (1013 hPa)
Smeltepunkt: 1455 °C
Tetthet: 8,9 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011137
CAS-nummer: 7440-02-0
EINECS: 231-111-4
FN: 3089
ADR: 4.1,II

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) insoluble
Melting Point 1453 °C
Molar Mass 58.69 g/mol
Bulk Density 1600 - 2600 kg/m3
Boiling Point 2832 °C
Density 8.90 g/cm3 (25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR