Tinn(II)sulfat, Sigma-Aldrich®

Leverandør: Merck

Synonymer: Tin(II) sulphate, Tin sulphate, Stannous sulphate

M-Clarity™ quality level = MQ200

1.12416.0250 1.12416.1000
1.12416.0250EA 955 NOK
1.12416.0250 1.12416.1000
Tinn(II)sulfat, Sigma-Aldrich®
Tinn(II)sulfat
Formel: O₄SSn
Tetthet: 4,15 g/cm³ (20 °C)
MDL nummer: MFCD00011246
CAS-nummer: 7488-55-3
EINECS: 231-302-2
Merck Indeks: 12,08947

Bestill nå

Spesifikasjon Testresultater

Solubility (20 °C) soluble
Molar Mass 214.77 g/mol
Bulk Density 800 kg/m3
pH 1.6 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR