Lactalbumin hydrolysate

Leverandør: Merck

1.12523.5000
1.12523.5000EA 16930 NOK
1.12523.5000
Lactalbumin hydrolysate
Proteiner

CAS No.: 68458-87-7
Physical Form: Powder
Appearance: Tan
MDL No.: MFCD00147378


Applications: A solubilized enzymatic digest of lactalbumin

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR