Spectromelt® C 20 grinding and tabletting aid, for X-ray analysis, Supelco®
Leverandør: Merck
M-Clarity™ quality level = MQ100
1.13934.5000
1.13934.5000EA 8650 NOK
1.13934.5000
Spectromelt® C 20 grinding and tabletting aid, for X-ray analysis, Supelco®
Spectromelt® C 20
Bestill nå
Spesifikasjon Testresultater
Identity (IR-spectrum) passes test
Al (Aluminium) ≤ 0.001
Ba (Barium) ≤ 0.001
Ca (Calcium) ≤ 0.001
Co (Cobalt) ≤ 0.001
Cr (Chromium) ≤ 0.001
Cu (Copper) ≤ 0.001
Fe (Iron) ≤ 0.001
K (Potassium) ≤ 0.001
Mn (Manganese) ≤ 0.001
Mo (Molybdenum) ≤ 0.001
Ni (Nickel) ≤ 0.001
Pb (Lead) ≤ 0.001
Si (Silicon) ≤ 0.005
Sr (Strontium) ≤ 0.001
Ti (Titanium) ≤ 0.001
V (Vanadium) ≤ 0.001