Sett for fargesammenlikning, MColortest™, Supelco®
Leverandør: Merck
1.14417.0001 1.14429.0001 1.14798.0001 1.18457.0002
1.14417.0001EA 1980 NOK
1.14417.0001 1.14429.0001 1.14798.0001 1.18457.0002
Sett for fargesammenlikning, MColortest™, Supelco®
Miljø testkit
Raske tester og visuell fargesammenlikning.
  • Resultater i løpet av 7 minutter
  • Enkel å bruke
  • Bruksområdeorienterte måleområder
  • Bærbar
  • Det finnes et utvalg av rimelige påfyllingspakker (RP)
MQuant® with colour card comparator
By using cuvettes with a long light path, the MQuant® (MCC) test kits are especially suited for measuring low ion concentrations (some as low as 0,005 mg/l). These test kits are especially appropriate for analysing drinking water, spring water, purified and mineral water, as well as cooling and boiler water.
MQuant® with colour disc comparator MCD
Using the comparator disc, these test kits are especially suitable for the analysis of turbid and coloured waters. The comparator disc is a practical and robust colour comparison device that provides reliable measurements, even in humid environments. Ideal for samples of medium to high concentration in applications ranging from testing bottled water to wastewater and other industrial water testing.
Leveringsinformasjon: Alle testsettene inneholder alle de nødvendige reagensene, kyvetter, fargesammenlikningskart eller komparatorhjul og en omfattende brukerveiledning.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
M-Clarity quality level MQ100