Sett for fargesammenlikning, Kobber MColortest™, Supelco®
Leverandør: Merck
1.18459.0002 1.14765.0001
1.18459.0002EA 2590 NOK
1.18459.0002 1.14765.0001
Sett for fargesammenlikning, Kobber MColortest™, Supelco®
Miljø testkit
Raske tester og visuell fargesammenlikning.
  • Resultater i løpet av fem minutter
  • Enkel å bruke
  • Bruksområdeorienterte måleområder
  • Bærbar
  • Det finnes et utvalg av rimelige påfyllingspakker (RP)
MQuant® with colour disc comparator
Using the comparator disc, these test kits are especially suitable for the analysis of turbid and coloured waters. The comparator disc is a practical and robust colour comparison device that provides reliable measurements, even in humid environments. Ideal for samples of medium to high concentration in applications ranging from testing bottled water to wastewater and other industrial water testing.
RP= Refill pack.
Leveringsinformasjon: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
M-Clarity quality level MQ100