Saltsyre 36%, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ100

1.15186.1000
1.15186.1000EA 1050 NOK
1.15186.1000
Saltsyre 36%, Supelco®
Saltsyre
Formel: HCl
Kokepunkt: 110 °C (1013 hPa)
Smeltepunkt: –30 °C
Tetthet: 1,18 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
FN: 1789
ADR: 8,

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR