TBX (Tryptone Bile X‑glucuronide) agar, Chromocult®
Leverandør: Merck
1.16122.0500
1.16122.0500EA 5310 NOK
1.16122.0500
TBX (Tryptone Bile X‑glucuronide) agar, Chromocult®
Mikrobiologiske medier Media, dehydrert for mikrobiologi
For the enumeration, detection and identification of β-glucuronidase positive Escherichia coli from food and feeding stuff as well as from environmental samples in the area of food production and food handling.
Sertifiseringer: Complies with the specifications given by ISO 16649 part 1 and part 2 and with those given by EN ISO 16649-3.