Reflectoquant®-teststrips, Supelco®
Leverandør: Merck
1.16730.0001
1.16730.0001EA 560 NOK
1.16730.0001
Reflectoquant®-teststrips, Supelco®
Miljø testkit
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly