Reflectoquant®-teststrips, Supelco®
Leverandør: Merck
1.16974.0001 1.16731.0001 1.17968.0001
1.16974.0001EA 665 NOK
1.16974.0001 1.16731.0001 1.17968.0001
Reflectoquant®-teststrips, Supelco®
Miljø testkit
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly
This test is suited for the determination of inorganic peroxides in aqueous solutions. Furthermore, it can be used in disinfecting and rinsing solutions (e.g., in food technology and laundries) and in swimming pool water.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
M-Clarity quality level MQ100