Dimetylsulfoksid, EMPLURA®, Supelco®

Leverandør: Merck

Synonymer: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

M-Clarity™ quality level = MQ200

1.16743.1000 1.16743.6025
1.16743.1000EA 757 NOK
1.16743.1000 1.16743.6025
Dimetylsulfoksid, EMPLURA®, Supelco®
Dimetylsulfoksid
Formel: (CH₃)₂SO
Kokepunkt: 189 °C (1013 hPa)
Tetthet: 1,101 g/cm³ (20 °C)
Flash punkt: 87 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002089
CAS-nummer: 67-68-5
EINECS: 200-664-3
Merck Indeks: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR