Eosin Y 1% alcoholic solution counterstain in H&E, for histology

Leverandør: Merck
Danger

Synonymer: Solvent Red 43, Field's Stain B, Eosin 225, 2,4,5,7-Tetrabromofluorescein disodium salt, Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate, Disodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), Tetrabromofluorescein disodium salt, 2′,4′,5′,7′-Tetrabromofluorescein disodium salt, Eosin Y disodium salt

1.17081.1000
1.17081.1000EA 1660 NOK
1.17081.1000
Eosin Y 1% alcoholic solution counterstain in H&E, for histology
Eosin Y

The hematoxylin and eosin (H&E) staining method is the method most frequently used for the staining of histology material of human origin.


This Eosin Y solution is a 1% alcoholic staining solution, that can be used e. g. for the routine staining of cryo sections. 1L of the solution will stain up to 1500 slides. It is an IVD / CE registered & certified product that can be applied in clinical diagnostics due to its excellent quality and reliable dye concentration

Formel: C₂₀H₆Br₄Na₂O₅
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005040
CAS-nummer: 17372-87-1
FN: 1170
ADR: 3,
IVDLogo

Bestill nå

Spesifikasjon Testresultater

Suitability for microscopy (Tissue section) passes test
Nuclei dark blue to dark violet
Cytoplasm pink to red
intercellular substances pink to red
Erythrocytes yellow to orange

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR