Air sampler, MAS-100 VF
Leverandør: Merck
1.17103.0001
1.17103.0001EA 0 NOK
1.17103.0001
Air sampler, MAS-100 VF
Prøvetakere Luftprøvetakere
The MAS-100 VF® uses standard Petri dishes for monitoring the quality of your environment. Its electronic speed control maintains an accurate flow rate. The handle enables the sampler to be mounted onto an optional tripod, thereby allowing testing at different angles for fixed applications. The perforated lid is the same as on the standard MAS-100® sampler.
  • Programmable delay before sampling starts and pre-selected volumes, individually adjustable
  • Graphic illuminated display and menu guided navigation with up and down arrows and direct menu selection