Capping A (Tetrahydrofurane:2,6-Lutidine:Acetic anhydride) (8:1:1) v/v/v for DNA/RNA synthesis
Leverandør: Merck
Danger
M-Clarity™ quality level = MQ200
1.18603.0450
1.18603.0450EA 1050 NOK
1.18603.0450
Capping A (Tetrahydrofurane:2,6-Lutidine:Acetic anhydride) (8:1:1) v/v/v for DNA/RNA synthesis
Capping Reagent
Flash punkt: 4 °C
FN: 2924
ADR: 3 (8)