Digital anemometer for the calibration of MAS-100 air samplers
Leverandør: Merck
1.19153.0001
1.19153.0001EA 0 NOK
1.19153.0001
Digital anemometer for the calibration of MAS-100 air samplers
Anemometere
MAS-100 air samplers are designed and manufactured to the most exacting standards. Ensuring compliant operation requires regular verification of the air flow of the instruments. This is because the air flow determines the impaction speed of the microbe-carrying particles from the air onto the agar surface and thus significantly influences the collection efficiency of your microbial air sampler. Regular checking of the air flow is therefore indispensable for your measurement reliability. 
  • The MAS-100 Regulus digital anemometer calibrates the following microbial air samplers