Nikotinamid-adenin dinukleotid (oksidert form), Millipore®

Leverandør: Merck

Synonymer: Nadide, NAD+, Nicotinamide adenine dinucleotide, oxidized

M-Clarity™ quality level = MQ200

1.24542.0001 1.24542.0005 1.24542.0025
1.24542.0001EA 682 NOK
1.24542.0001 1.24542.0005 1.24542.0025
Nikotinamid-adenin dinukleotid (oksidert form), Millipore®
Nikotinamid-adenin dinukleotid (oksidert form)
Formel: C₂₁H₂₇N₇O₁₄P₂
Smeltepunkt: 140 to 142 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00036253
CAS-nummer: 53-84-9
EINECS: 200-184-4
Merck Indeks: 13,06370

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR