AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride), Sigma-Aldrich®

Leverandør: Merck
Warning

Synonymer: 4-(2-Aminoethyl)benzenesulphonyl fluoride hydrochloride, AEBSF hydrochloride

M-Clarity™ quality level = MQ100

1.24839.0100 1.24839.0500
1.24839.0100EA 970 NOK
1.24839.0100 1.24839.0500
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride), Sigma-Aldrich®
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
Formel: C₈H₁₁ClFNO₂S
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00132962
CAS-nummer: 30827-99-7
FN: 3261
ADR: 8,II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR