Hygienekontroll, HY-LiTE® 2, ATP luminometer
Leverandør: Merck
1.30100.0001
1.30100.0001EA 0 NOK
1.30100.0001
Hygienekontroll, HY-LiTE® 2, ATP luminometer
Overvåkningssystemer Hygieneovervåkningssystemer
Rapid, quantitative hygiene monitoring in liquids and on surfaces with ATP-bioluminescence HY-LiTE® 2, contained in a convenient lightweight shoulder bag with data logging and TREND2 software for advanced data management. The HY-LiTE® TREND2 software can be used to compile HACCP plans and individual limits. Graphs showing results make them easy to evaluate and interpret. HY-LiTE® is already used in a wide variety of production and processing fields, like the food industry, packing industry, services (cleaning of buildings), community catering and large scale kitchens, drinks manufacturers,More Product Information