HPLC columns, LiChroCART®, Superspher® 60 RP-select B
Leverandør: Merck
1.50173.7141 1.50178.7141 1.50179.7141 1.50195.7141 1.50228.7141 1.50229.7141 1.50234.7141 1.50236.7141 1.50302.7141 1.51431.7141 1.51432.7141 1.51442.7141 1.51443.7141 1.51444.7141 1.51448.7141 1.50170.7141 1.50201.7141 1.50175.7141
1.50173.7141EA 0 NOK
1.50173.7141 1.50178.7141 1.50179.7141 1.50195.7141 1.50228.7141 1.50229.7141 1.50234.7141 1.50236.7141 1.50302.7141 1.51431.7141 1.51432.7141 1.51442.7141 1.51443.7141 1.51444.7141 1.51448.7141 1.50170.7141 1.50201.7141 1.50175.7141
HPLC columns, LiChroCART®, Superspher® 60 RP-select B
Kromatografikolonner
Stainless steel LiChroCART® cartridges are available with various types of Superspher® sorbents.
These LiChroCART® cartridges require part number 1.51486.0001 manu-CART® cartridge column holder, which can be used to hold one cartridge column with or without a  4×4 mm guard column. LiChroCART® columns 250×10 mm require part number 1.51419.0001 manu-CART® 10. The short LiChroCART® columns can be ordered as a set including the corresponding cartridge holder and one cartridge, or as a pack of 3 cartridges without cartridge holder. The separate part numbers for the cartridge are asMore Product Information