HPLC columns, Hibar®, LiChrospher® 100 CN
Leverandør: Merck
1.00423.7070 1.00424.7070 1.50009.7070 1.50012.7070 1.50013.7070 1.50182.7070 1.50183.7070 1.00424.7071 1.50182.7071 1.50009.7071 1.50183.7071 1.50013.7071
1.00423.7070EA 12200 NOK
1.00423.7070 1.00424.7070 1.50009.7070 1.50012.7070 1.50013.7070 1.50182.7070 1.50183.7070 1.00424.7071 1.50182.7071 1.50009.7071 1.50183.7071 1.50013.7071
HPLC columns, Hibar®, LiChrospher® 100 CN
Kromatografikolonner
Stainless steel Hibar® columns are complete with endfittings. These columns are available with various types of LiChrospher® sorbents.
When using a guard column with a Hibar® RT column, we recommend part number 1.51487.0001 guard column cartridge holder for 4×4 mm guard column cartridges LiChroCART®.