HPLC columns, SeQuant® ZIC®-pHILIC
Leverandør: Merck
1.50437.0001 1.50459.0001 1.50460.0001 1.50461.0001 1.50462.0001 1.50463.0001 1.50464.0001 1.50438.0001
1.50437.0001EA 4625 NOK
1.50437.0001 1.50459.0001 1.50460.0001 1.50461.0001 1.50462.0001 1.50463.0001 1.50464.0001 1.50438.0001
HPLC columns, SeQuant® ZIC®-pHILIC
Kromatografikolonner
ZIC®-pHILIC has the same stationary phase as ZIC®-HILIC, but is instead based on a polymeric particle for enhanced pH stability. The lack of silica practically nullifies background in high sensitivity detectors such as charged aerosol used for determination of organic and inorganic ions.
  • Solves extra challenging separations of polar and hydrophilic compounds
  • Enhanced selectivity optimisation potential due to very broad pH application range
  • Extremely low background with high sensitivity charged aerosol evaporative ion detectors