HPLC columns, Chromolith® Phenyl
Leverandør: Merck
1.52059.0001
1.52059.0001EA 3725 NOK
1.52059.0001
HPLC columns, Chromolith® Phenyl
Kromatografikolonner
Chromolith® Phenyl HPLC columns offer greater selectivity towards aromatic ring-containing compounds than standard alkyl phases. These columns are ideal for the separation of aromatic compounds, flavonoids, fatty acids, PAH, preservatives, purines and pyrimidines.
  • High throughput at high flow rates
  • Possibility of flow gradients
  • Added column performance by column coupling
  • Rigid monolithic structure for longer lifetime
  • Less matrix-sensitivity
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Karbonbelastning
Overflate areal