Vann med 0,1% (v/v) Maursyre ( metansyre), LiChrosolv®, hypergrade LC-MS, Supelco®
Leverandør: Merck
MQ100
1.59013.2500
1.59013.2500EA 815 NOK
1.59013.2500
Vann med 0,1% (v/v) Maursyre ( metansyre), LiChrosolv®, hypergrade LC-MS, Supelco®
Vann med 0,1% (v/v) Maursyre ( metansyre)
LiChrosolv® hypergrade is accurately tested for LC-MS suitability, meeting all the requirements of modern LC-MS ionisation methods (ESI / APCI - positive and negative mode).
Thanks to their low level of ionic background and low ion suppression, LiChrosolv® hypergrade solvents provide high reproducibility and high ionisation efficiency. The packaging material has been improved to perfectly meet LC-MS quality requirements.