Salpetersyre 25%, Titripur® for argentometric titration, Supelco®

Leverandør: Merck
Danger

Synonymer: Nitric acid solution

M-Clarity™ quality level = MQ100

1.60317.0500
1.60317.0500EA 420 NOK
1.60317.0500
Salpetersyre 25%, Titripur® for argentometric titration, Supelco®
Salpetersyre
Formel: HNO₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011349
CAS-nummer: 7697-37-2
FN: 2031
ADR: 8,

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR