Natriumhydrogenkarbonat FCC, E500, SAFC®

Leverandør: MERCK PRODUCTION CHEMICALS

1.72577.9025
1.72577.9025EA 0 NOK
1.72577.9025
Natriumhydrogenkarbonat FCC, E500, SAFC®
Natriumhydrogenkarbonat
Formel: NaHCO₃
Smeltepunkt: 270 °C
Tetthet: 2,159 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003528
CAS-nummer: 144-55-8
EINECS: 205-633-8
Merck Indeks: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR