Human Serum AB (Off the Clot)
Leverandør: GEMINI BIO-PRODUCTS
100-318-100
100-318-100EA 9375 NOK
100-318-100
Human Serum AB (Off the Clot)
Serum
The off-the-clot human male AB serum is collected from blood that is allowed to coagulate naturally after collection, so there are no anticoagulants or preservatives.
  • Recommended for pre-clinical through clinical workflows
  • 0,1 µm sterile-filtered
Off the clot human serum AB is collected from normal healthy male donors at FDA-licensed facilities in the United States. As a natural off the clot product, this material contains no anticoagulants/preservatives. All donor units are tested for viral markers and found to be non-reactive.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Lagringstemp Store frozen at −20…−10 °C