GemCell Plus™ U.S. Human Serum AB - Xeno-Free
Leverandør: GEMINI BIO-PRODUCTS
100-912-100
100-912-100EA 8650 NOK
100-912-100
GemCell Plus™ U.S. Human Serum AB - Xeno-Free
Serum
GemCell Plus™ Xeno-Free Human Serum is collected from healthy male donors of the AB serotype at FDA-licensed facilities in the United States. This material goes through additional rigorous testing and contains therapeutic grade human thrombin. All donor units are tested for viral markers and found to be non-reactive.
  • Developed without the use of animal material
  • Gamma irradiation available