Acarbose, Reference Standard
Leverandør: USP
1000521
1000521.EA 5325 NOK
1000521.
Acarbose, Reference Standard
Standarder Organiske standarder
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Sertifiseringer: US Pharmacopea Reference Standard
NB: Cold Shipment required
Bestill nå
SPESIFIKASJONER
Beskrivelse Acarbose
Environmentally Preferable
CAS-nr 56180-94-0
Formel C₂₅H₄₃NO₁₈
Lagringsforhold Refigerator