Acarbose System Suitability Mixtures, Reference Standard
Leverandør: USP
1000532
1000532.EA 13700 NOK
1000532.
Acarbose System Suitability Mixtures, Reference Standard
Standarder Organiske standarder
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
A mixture containing acarbose and impurities A, B, C, D, E, F and G
Sertifiseringer: US Pharmacopea Reference Standard
NB: Cold Shipment required