L(+)-Lactic acid 88.0-92.0%, AnalaR NORMAPUR® analytisk reagens

Leverandør: VWR Chemicals
Danger

Synonymer: (S)-Lactic acid, L-Lactic acid, (S)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropionic acid, (S)-(+)-Lactic acid

101384QEA 1150 NOK
101384Q
L(+)-Lactic acid 88.0-92.0%, AnalaR NORMAPUR® analytisk reagens
L(+)-Lactic acid
Formel: C₃H₆O₃
MW: 90,08 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064266
CAS-nummer: 79-33-4
EINECS: 200-018-0
Merck Indeks: 13,05351

Bestill nå

Spesifikasjon Testresultater

Assay 88.0 - 92.0 %
IR Spectrum Passes test
Reducing substances Passes test
Volatile fatty acids Passes test
Colouration Max. 20 APHA
Relative density 1.200 - 1.210
Ignition residue (SO4) Max. 100 ppm
Cl (Chloride) Max. 10 ppm
SO4 (Sulphate) Max. 50 ppm
As (Arsenic) Max. 0.1 ppm
Ca (Calcium) Max. 0.02 %
Cu (Copper) Max. 1 ppm
Fe (Iron) Max. 3 ppm
Pb (Lead) Max. 1 ppm
Zn (Zinc) Max. 10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR