Etylenglykol ≥99%, Reagent Grade

Leverandør: Honeywell Chemicals
Warning

Synonymer: Ethane-1,2-diol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, Glycol

102466-2.5L 102466-20L 102466-30KG 102466-5L 102466-1L 102466-225KG
102466-2.5LEA 2675 NOK
102466-2.5L 102466-20L 102466-30KG 102466-5L 102466-1L 102466-225KG
Etylenglykol ≥99%, Reagent Grade
Etylenglykol

  • ≥95% for general laboratory use.

Formel: HOCH₂CH₂OH
MW: 62.07 g/mol
Kokepunkt: 197 °C
Smeltepunkt: –12 °C
Tetthet: 1.11 g/cm³ (20 °C)
Flash punkt: 111 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002885
CAS-nummer: 107-21-1
EINECS: 203-473-3
REACH: 01-2119456816-28
Merck Indeks: 13,03832
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d1c97392851ba184f445f6ee912f84631f98d9&vid=2002300f174929a030d66d9f1394eb6bee89b7dc&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Refractive Index n20/D 1.431 (lit.)
Assay (GC) Min. 99.0 %
IR spectrum complying
APHA Max. 10
Vapor Density 2.1 (vs air)
Explosion Limit 15.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR