Niobium ≥99.8% (metals basis), metalltråd, Ø 1.0 mm (0.04 in)

Leverandør: Thermo Fisher Scientific

10264.G1 10264.G5 10264.G9
10264.G1EA 720 NOK
10264.G1 10264.G5 10264.G9
Niobium ≥99.8% (metals basis), metalltråd, Ø 1.0 mm (0.04 in)
Niobium

Purity: 99.8% CAS No.: 7440-03-1 Physical Form: ≈6.7g/m Odour: Odourless MDL No.: MFCD00011126

Formel: Nb
Kokepunkt: 3700 °C (1013 hPa)
Smeltepunkt: 2468 °C
Tetthet: 8,57 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011126
CAS-nummer: 7440-03-1
EINECS: 231-113-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR