Diammoniumhydrogenortofosfat P₂O₅ ≥ 53%, tech.

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate

10301.A1 10301.A7
10301.A1EA 825 NOK
10301.A1 10301.A7
Diammoniumhydrogenortofosfat P₂O₅ ≥ 53%, tech.
Diammoniumhydrogenortofosfat

CAS No.: 7783-28-0 Molecular Formula: (NH4)2HPO4 Formula Weight: 132.06 Physical Form: -100 Mesh Powder Density: 1.619 Appearance: White Melting Point: 155° dec. Solubility: Freely soluble in water. Insoluble in alcohol, acetone Merck Reference: 14,542 MDL No.: MFCD00010891


Corrosion inhibitors, in fermentation, as flux for soldering tin, copper, brass, and zinc

Formel: (NH₄)₂HPO₄
MW: 132.06 g/mol
Smeltepunkt: 155 °C (dec.)
Tetthet: 1.619
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010891
CAS-nummer: 7783-28-0
EINECS: 231-987-8
REACH: 01-2119490974-22
Merck Indeks: 14,542

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR