tri-Ammoniumcitrat 98.0-101.0%, AnalaR NORMAPUR® analytical reagent

Leverandør: VWR Chemicals
Warning

Synonymer: 2-Hydroxy-1,2,3-propanetricarboxylic acid triammonium salt, Ammonium citrate tribasic, Citric acid triammonium salt, Triammonium 2-hydroxypropane-1,2,3-tricarboxylate

103013AEA 870 NOK
103013A
tri-Ammoniumcitrat 98.0-101.0%, AnalaR NORMAPUR® analytical reagent
tri-Ammoniumcitrat
Formel: C₆H₁₇N₃O₇
MW: 243.22 g/mol
Smeltepunkt: 185
Tetthet: 1 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00036406
CAS-nummer: 3458-72-8
EINECS: 222-394-5

Bestill nå

Spesifikasjon Testresultater

Assay 98.0 - 101.0 %
Identification Passes test
Reducing substances Passes test
pH (20°C; 5 %) 6.0 - 7.5
Ignition residue (SO4) Max. 50 ppm
Insolubility in water Max. 50 ppm
Cl (Chloride) Max. 10 ppm
Cu (Copper) Max. 1 ppm
Fe (Iron) Max. 5 ppm
K (Potassium) Max. 30 ppm
Na (Sodium) Max. 40 ppm
Pb (Lead) Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR